Joint clamps, stainless steel
Supplier: HWS LABORTECHNIK
Certificates
About this item
Stainless steel wire.
- High clamping force for a secure hold of the ground joints
- High resistance to aggressive media
- Very good thermal and mechanical resistance
Stainless steel wire.