Order Entry
Germany
ContactUsLinkComponent
VWR®, Clamps, Four Pronged
VWR®, Clamps, Four Pronged
Catalog # 241-0029
Supplier:  VWR Collection
undefined
VWR®, Clamps, Four Pronged
Catalog # 241-0029
Supplier:  VWR Collection

Specifications

  • Spanning width:
    35 - 100 mm
  • Pk:
    2

Specifications

About this item

Stainless steel.

  • Robust design, clamp rod diameter 12 mm, 140 mm long
  • Two easy action knurled screws for adjusting the clamping fingers independently of each other