VWR®, Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Certificates
About this item
PS, sterile. Calibrated, disposable inoculating loops made from flexible or rigid plastic that cater for microbiologist's different applications and methods.
- Colour coded for easy product identification
- Conveniently packed in anti-roll tubes with push-on resealable cap
- Fast, easy and secure access to the loops, without the need for a separate holder
- Loops have an ultra-smooth surface for easy streaking and length of 210 mm
Delivery: Calibration certificate supplied with every pack.