Cell scrapers, Falcon®
Supplier: Corning
Certificates
About this item
PS handle, TPE blade. Designed to provide maximum accessibility to the growth surfaces of a variety of culture vessels.
- Cross-ribbed PS handle provides greater rigidity, to ensure better control while scraping cells
- Highly compliant TPE blade pivots to provide multiple angles to remove cells from the entire growth surface
Ordering information: Supplied individually wrapped in peel-open packages.
Specifications
- Catalog No:
- 734-0386
- 734-0387
- 734-1111
- 734-0385
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa