Order Entry
Germany
ContactUsLinkComponent
Transport carriers, Bel-Art Products, a part of SP
Transport carriers, Bel-Art Products, a part of SP
Catalog # 215-0170
undefined
Transport carriers, Bel-Art Products, a part of SP
Catalog # 215-0170

Specifications

  • Description:
    Multipurpose carrier
  • Seq:
    1
  • Pk:
    1

Specifications

About this item

PP.

  • Robust design, strong walls
  • Comfortable round carrying handle
  • Insert for six compartments (optional)

These carriers are suitable for transporting bottles, samples, tools and utensils.

W×D×H: 360×180×150 mm

Ø (hole): 74 mm

Colour: white