Burette Covers
Catalog # 612-9951
Supplier: Brand
Specifications
- Description:Burette covers, 20Ø×30H mm
- Pk:10
Specifications
About this item
Made of PP. Ribbed inside to prevent suction attachment during titration.
- Ribbed inside to prevent suction attachment during titration
ØxH: 20x30 mm