Stainless Steel Buckets
Supplier: RSG
Jump to:
About this item
Stainless steel, rust-proof, polished finish.
- Very good chemical and thermal resistance
- High quality, very stable
- Optional matching lids are available
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.
4 Options Available Below
Product Details & Documents
Stainless steel, rust-proof, polished finish.
- Very good chemical and thermal resistance
- High quality, very stable
- Optional matching lids are available
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.
Product Family Options
Product Information
Lids for buckets
Product Information
- Catalog #ForDescriptionAvailabilityPrice