Order Entry
Germany
ContactUsLinkComponent
Labtop cooler −20 °C, Nunc™
Catalog # 391-7635
   Low Manufacturing Impact
undefined
Labtop cooler −20 °C, Nunc™
Catalog # 391-7635
   Low Manufacturing Impact

Specifications

  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    1

Specifications

About this item

Impact resistant PC.

  • Alphanumeric printed grid on the transparent one way keyed lid
  • Holds 20 CryoTubes™ (1,0 and 1,8 ml), inserts are included to accommodate 0,2 to 0,5 ml microtubes
  • Stackable
  • With locking handle

Protects enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench. Maintains freezer temperature during the frost-free cycle, power failure and accidental door opening.