Orders LinkContactUsLinkComponent
Quick Chill™ Cooling unit, Nalgene®
Quick Chill™ Cooling unit, Nalgene®
Catalog # 479-5160
   Low Manufacturing Impact
Quick Chill™ Cooling unit, Nalgene®
Catalog # 479-5160
   Low Manufacturing Impact
Quick Chill™ Cooling unit, Nalgene®
Quick Chill™ Cooling unit, Nalgene®
Price
163,00 €
Sale
Valid until 30/06/2026
Log in for availability
1 * 1 ST
Product Details & Documents
PC, non-toxic insulating solution.
  • No direct contact between tubes and insulating solution so labels and markings won't come off
  • Stackable and have non-slip rubber feet
For rapid chilling of biological samples. Can be used repeatedly for up to 45 minutes. It will chill up to 2 ml of alcohol-based solutions to −20 °C in less than 3 minutes or up to 2 ml of water-based solutions to 0 °C in less than 4 minutes.
For 12 (3×4) microtubes of 1,5 or 0,5 ml.
W×D×H: 151×108×125 mm
Specifications
  • Cat. No.:
    479-5160
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact