Safety wash bottles, narrow mouth, printed, VITsafe™
Supplier: VITLAB
Certificates
About this item
LDPE, with VENT CAP threaded connection.
- Colour coded contents with hazard symbols and descriptions of hazards as required by the Hazardous Substance Ordinance
- Good chemical resistance
- Good thermal resistance
VENT CAP works on the principle of gas/vapour permeability. The screw cap prevents any build-up of vapour pressure above the contents. This prevents the risk of solvent dripping or leaking.
Caution: Note: Washbottle 'Hexan' is not suitable for n-Hexane.