Safety wash bottles, narrow/wide neck, with printed name
Supplier: VITLAB
Certificates
About this item
LDPE, translucent with PP screw cap.
- Good chemical resistance
- Heat resistant up to 80 °C (up to 90 °C for brief periods)
Coloured inscription of contents in German, English and French.