Safety waste funnels, Nalgene®
HDPE funnels and covers, with PP closure and funnel adapter, PTFE vent membrane.
- Vent system helps reduce fumes and splashing
- Removable neck screen traps large particles
- Available in 10" Ø funnel with 83B closure and 5¹/₂" funnel with 38 to 430 closure
For solvent, chemical and biological waste. Features the easy snap safety latch to minimise spillage if accidentally tipped over and a larger diameter funnel for easy pouring.
- Catalog No:
- 119-3034
- 119-3040
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement