Test tube baskets
PP.
- Very good chemical resistance
- Autoclavable at 135 °C
- Highly break-resistant
- Stackable space saving design
These tests tubes baskets are suitable for rinsing, sterilising, transporting and storing glassware.
Colour: White
PP.
These tests tubes baskets are suitable for rinsing, sterilising, transporting and storing glassware.
Colour: White