Autoclavable Bags
Supplier: Greiner Bio-One
Certificates
About this item
These bags are designed specifically for the disposal of contaminated laboratory waste, that first requires steam sterilisation.
- Bags for autoclaving and oven - resistant up to 160 ° C in an oven and 134 ° C in an autoclave
- Suitable for hot air sterilisers