Autoclave Bags
Supplier: Greiner Bio-One
Certificates
About this item
These disposal bags are used for storing contaminated petri dishes or test tubes.
- Suitable for autoclaving by wet steam
- Specially designed to eliminate contaminated waste in the laboratory
PP, transparent, thickness: 50 µm