ContactUsLinkComponent
Anti-static spray, Antistatik 100
Anti-static spray, Antistatik 100
Catalog # 291-1240
undefined
Anti-static spray, Antistatik 100
Catalog # 291-1240
Specifications
  • Description:
    Antistatic Spray
  • Pk:
    200 mL
Specifications
About this item
ANTISTATIK 100 forms an invisible protective film against electrostatic charges. The antistatic solution with cleaning alcohol removes dirt and offers long-lasting protection against static electricity. ANTISTATIK 100 cleans and protects plastic housings, textile surfaces, screens, acrylic glass covers or packaging films.