Seals for flanged connections
Catalog # 201-0126
Supplier: Lenz Laborglas GmbH & CO.KG
Specifications
- For:Ball or flat flanges KF/PF 15
- Colour:White
- Description:Seals for flanged connections
- Material:PTFE
- Pk:1
Specifications
About this item
PTFE, white.
- Simple easy assembly and disassembly
- Very good chemical resistance
- Very good thermal resistance
For impermeable ball or flat flange connections.