Order Entry
Germany
ContactUsLinkComponent
Pararosaniline hydrochloride ≥85% (by HPLC), Sigma-Aldrich®
Health
Pararosaniline hydrochloride ≥85% (by HPLC), Sigma-Aldrich®
Catalog # P3750-25G
CAS Number:  
undefined
Health
Pararosaniline hydrochloride ≥85% (by HPLC), Sigma-Aldrich®
Catalog # P3750-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Colour):
    Green to Very Dark Green
  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Solubility (Turbidity)
    Clear to Opaque
    c = 1 mg/ml; Ethanol:
    Conforms
  • 59.9 - 81.0 %:
    Conforms
  • 11.0 - 14.9 %:
    Carbon
  • 543 - 549 nm:
    Nitrogen
  • ≥80000:
    Wavelength
    c = 0.003 g/L; 50% Ethanol
  • 286 - 292 nm:
    Extinction Coefficient
  • ≥17000:
    Wavelength
  • ≥85%:
    Extinction Coefficient
  • undefined:
    Purity (HPLC)

Specification Test Results

About this item

 :
 :323.82 g/mol
Melting Pt:268...270 °C
 :M-Clarity™ quality level = MQ200
EINECS:209-321-2
 :GHS08
Storage Temperature:Room Temperature
CAS Number:569-61-9
Formula:C₁₉H₁₈ClN₃
MDL Number:MFCD00001657