Sodium diphenylamine-4-sulphonate, Millipore®
Catalog # 1.03590.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:820 g/l (20 °C)
- Melting Point:200 °C
- Molar Mass:271.26 g/mol
- Bulk Density:160 kg/m3
- pH:5.7 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:271.27 g/mol
Merck Index:13,07397
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:228-165-6
CAS Number:6152-67-6
Formula:C₁₂H₁₀NNaO₃S
MDL Number:MFCD00036468