Sodium diphenylamine-4-sulphonate ACS
:6152-67-6
:
:C₁₂H₁₀NNaO₃S
:MFCD00036468
:271.27 g/mol
:13,07397
:228-165-6
:GHS07
:Ambient
- Appearance:Off-white to beige to light grey Crystalline powder
- Infrared spectrum:Conforms
- Sensitivity:(as indicator) passes test