Order Entry
Germany
ContactUsLinkComponent
Tritolyl phosphate (mixture of isomers)
Irritant
Environment
Tritolyl phosphate (mixture of isomers)
Catalog # EHERC17800800
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Environment
Tritolyl phosphate (mixture of isomers)
Catalog # EHERC17800800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :368.37 g/mol
Merck Index:12,09892
Boiling Pt:420 °C (1013 hPa)
Melting Pt:–40 °C
Density:1.16 g/cm³ (20 °C)
EINECS:215-548-8
UN:2810
ADR:6.1,II
 :GHS07
 :GHS09
CAS Number:1330-78-5
Formula:C₂₁H₂₁O₄P
MDL Number:MFCD02102116