

Tritolyl phosphate (mixture of isomers) ≥98% ortho-isomers <1%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1330-78-5
:
Formula:C₂₁H₂₁O₄P
MDL Number:MFCD02102116
:368.37 g/mol
Merck Index:14,9763
Boiling Pt:265 °C (10 mmHg)
Melting Pt:–40 °C
Density:1.167
Flash Pt:250 °C (482 °F)
Storage Temperature:Ambient
EINECS:215-548-8
UN:2574
ADR:6.1,II
:GHS08
:GHS09