Carboxymethyl cellulose sodium salt, low viscosity, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9004-32-4
:
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472
Merck Index:12,01877
Melting Pt:300 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient