Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2-Methylthiazole

1
Catalog # WAKO326-44552-25G
Marketsource
Supplier:   Wako
2-Methylthiazole
Catalog # WAKO326-44552-25G
Supplier:  Wako
Marketsource
CAS Number:  3581-87-1
2-Methylthiazole

1

CAS Number:  3581-87-1
Price
Inquire For Price
Log in for availability
1 * 1 Pack.
Product Details & Documents
  • CAS Number:3581-87-1
  • Formula:C₄H₅NS
  • MDL Number:MFCD00053144
  • Molecular Weight:99.16 g/mol
  • EINECS:222-702-8
Specifications

Specifications

  • Pk:
    1

Specifications