Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Phthalide

Glass bottle for solids, 100 g
Catalog # MOLE25999254-100G
Irritant
Marketsource
Supplier:   Molekula
Phthalide
Catalog # MOLE25999254-100G
Supplier:  Molekula
Marketsource
CAS Number:  87-41-2
Phthalide

Glass bottle for solids, 100 g

CAS Number:  87-41-2
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Isobenzofuran-1(3H)-one
  • CAS Number:87-41-2
  • Formula:C₈H₆O₂
  • MDL Number:MFCD00005906
  • Molecular Weight:134.13 g/mol
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:73 °C
  • Density:1,1 g/cm³ (30 °C)
  • EINECS:201-744-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications