Order Entry
Germany
Orders LinkContactUsLinkComponent
Phthalide 99%

Phthalide 99%

Irritant
Phthalide 99%
Certificates
Phthalide 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Isobenzofuran-1(3H)-one
  • CAS Number:87-41-2
  • Formula:C₈H₆O₂
  • MDL Number:MFCD00005906
  • Molecular Weight:134.13 g/mol
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:70…73 °C
  • Density:1,1 g/cm³ (30 °C)
  • Flash Pt:152 °C
  • EINECS:201-744-0

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    70 to 73 °C
  • GC:
    ≥98.5 %
  • UV:
    at 230 nm A: ≤0.15
  • Color scale:
    ≤5 APHA
  • Residue after evaporation:
    ≤0.001 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,60 €
    21,60 €
    1 * 5 g
       21,60 €

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 41,00 €
    41,00 €
    1 * 100 g
       41,00 €

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 103,00 €
    103,00 €
    1 * 500 g
       103,00 €