Order Entry
Germany
Orders LinkContactUsLinkComponent
Thymolphthalein | Avantor

Thymolphthalein

Thymolphthalein
Certificates
Thymolphthalein

3 Options Available Below

Product Details & Documents

CAS No.: 125-20-2 Molecular Formula: C28H30O4 Formula Weight: 430.55 Melting Point: 251-253° Beilstein Reference: 359413 Merck Reference: 14,9401 MDL No.: MFCD00005909

Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:14,9401
  • Melting Pt:251…253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.