Ninhydrin solution R2 Reag. Ph. Eur. 1058305
Catalog # REGE1058305
Supplier: REAGECON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:178.14 g/mol
:GHS05
Storage Temperature:Ambient
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791