EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:67-42-5
:
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291
Merck Index:13,03560
Melting Pt:240…244 °C
:380.35 g/mol
Storage Temperature:Ambient
EINECS:200-651-2