Order Entry
Germany
ContactUsLinkComponent
VWR®, Karl Fischer NewGen Solvent-FI for volumetric two-component KF titration
VWR®, Karl Fischer NewGen Solvent-FI for volumetric two-component KF titration
Catalog # 88426.290
Supplier:  VWR Chemicals
New Product
CAS Number:  
undefined
VWR®, Karl Fischer NewGen Solvent-FI for volumetric two-component KF titration
Catalog # 88426.290
Supplier:  VWR Chemicals
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    NewGen Solvent-FI for volumetric two-component Karl Fischer titration
  • Seq:
    130

Specifications

About this item

Medium for volumetric two-component Karl Fischer titration (methanol-based), Free of imidazole (free of CMR substances)

  • free of imidazole
  • free of CMR substances

Specification:
Suitability (for KF-Titration): complying
Water content: 0.010 - 0.020% (v/v)