ContactUsLinkComponent
EZ-Fit™ Manifold
EZ-Fit™ Manifold
Catalog # MILFEZFITMIC03
Supplier:  Merck
EZ-Fit™ Manifold
Catalog # MILFEZFITMIC03
Supplier:  Merck
Specifications
  • Description:
    EZ-Fit™ Manifold, 3-place for Microfil® funnels and membranes
  • Material:
    Stainless steel
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
EZ-Fit™ Manifold for universal laboratory filtration.
  • Unique design for easy cleaning and decontamination
  • Different filtration heads for both reusable and disposable filtration devices
  • Quick-fit connectors for the vacuum tubing
  • Low height for ease of use in laminar flow hoods
  • Each component can be removed quickly and easily without any additional tools
  • A check valve is integrated into the tubing connection to prevent liquid back-flow from the vacuum system
  • The 2-way valve prevents the user from accidentally touching the filtration head while opening or closing it
The new EZ-Fit™ Manifold design eliminates sources of contamination that can cause false positive results: all internal areas are accessible and can be cleaned efficiently and all components can be autoclaved.