Order Entry
Germany
ContactUsLinkComponent
 
MultiScreen® Vacuum Manifold 96 Well
  736-0190
 :  Merck
undefined
MultiScreen® Vacuum Manifold 96 Well
  736-0190
 :  Merck

 

  • Description:
    Vacuum manifold
  • Height:
    With standard ring: 66 mm
    With deep well ring: 94 mm
  • Length:
    152 mm
  • Pk:
    1
  • Width:
    122 mm

 

 

96-well plate Vacuum Manifold for vaccum filtration.

Components: EPDM rubber gasket, EPDM rubber seal, nylon well ring (standard), Tygon® tubing (FEP-lined), brass socket (for vacuum control valve), high-density polyethylene base, high-density polyethylene well ring (deep), polypropylene valve (on/off), stainless steel grid (for support), steel case (for vacuum control valve), steel gauge (for vacuum).