ContactUsLinkComponent
MultiScreen® Collection Plates, 96 Well
MultiScreen® Collection Plates, 96 Well
Catalog # MILFMATRNPS50
Supplier:  Merck
undefined
MultiScreen® Collection Plates, 96 Well
Catalog # MILFMATRNPS50
Supplier:  Merck
Specifications
  • No. of wells:
    96
  • Sterile:
    -
  • Colour:
    Clear
  • Pk:
    50
  • Well volume:
    100 μl sample volume (per well)
Specifications
About this item
These 96 well collection plates are suitable for mammalian cell culture and parallel artificial membrane permeation assay (PAMPA).
  • Transport receiver plate
  • Clear
  • Non sterile
Made of polystyrene