ContactUsLinkComponent
Chemical Duty Pump Rebuild Kit
Chemical Duty Pump Rebuild Kit
Catalog # MILFWP61RBD00
Supplier:  Merck
undefined
Chemical Duty Pump Rebuild Kit
Catalog # MILFWP61RBD00
Supplier:  Merck
Specifications
  • Description:
    Chemical duty pump rebuild kit
  • Pk:
    600
Specifications
About this item
Delivery: This item includes parts for catalog numbers WP6111560, WP6122050, WP6110060. This item is a kit composed of 1 diaphragm hold-down plate assembly, 1 valve plate assembly, 1 pump head, 1 vacuum gauge, 1 pressure gauge, 4 head screws, 2 handle screws.