Order Entry
Germany
ContactUsLinkComponent
Dishwashing Detergent, ProCare Universal 77 A
Dishwashing Detergent, ProCare Universal 77 A
Catalog # MIEL12456030
Supplier:  MIELE
New Product
Dishwashing Detergent, ProCare Universal 77 A
Catalog # MIEL12456030
Supplier:  MIELE
New Product

Specifications

  • Description:
    Machine cleaner, ProCare Universal 77 A
  • Pk:
    500 g
  • Temperature range:
    5...35 °C

Specifications

About this item

Machine cleaner, alkaline for removing persistent starch deposits.

  • Removal of grease, oil and protein
  • Reduction in odours
  • Good cleaning capability
  • Very gentle on materials
  • High percentage of dispersants
  • Intensive cleaning agent containing surfactants
  • Strong emulsifying and dispersing effect
  • Specially developed for Miele Professional machines

Ingredients:
Phosphate
Oxygen-based bleaching agent
Phosphonates
Polycarboxylates
Non-ionic surfactants

Certifications: GHS/CLP compliant.