Specifications
- Colour:Green/grey
- Description:Megasonic planet goggles
- Lenses:Clear PC lens
- Pk:1
Specifications
About this item
The uvex megasonic’s revolutionary lens design has an exceptional field of vision to ensure the best possible visibility at all times – even under difficult conditions. The sporty, ergonomic design and ultra-comfortable fit means these goggles both protect and allow you to get on with your day.
- Functional ski-style goggles with panoramic view
- Metal free
- Frame colour: anthracite
- Headband: anthracite, jade
- PC lens: clear
- UV400 – 100% protection against harmful UV radiation to 400 nm
- Increased mechanical strength – certification BT (120 m/s, tested under extreme temperatures –5 °C, +55 °C)
Unique, frameless lens design with maximum optimised field of view for better hazard perception and increased safety. uvex protecting planet – mass-balanced PC lens, frame, headband and packaging partially made from recycled material. Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens.
Soft, adaptive face pad provides a universal fit for different head and facial shapes and a tight fit without pressure points. Generous internal volume and openings for side arms allow comfortable wear without pressure points, even for prescription spectacle wearers. Indirect ventilation system for increased air circulation. 30 mm-wide, length-adjustable fabric headband for optimum fit and individual adjustment.
Certification: W 166 34 BT CE – 2C-1.2 W1 BTKN CE