Disposable Particle Filtering Respirators, FFP1/FFP2/FFP3
About this item
Respirators in protection class FFP1, FFP2 and FFP3.
- Soft material edges for greater comfort and wearer acceptance
- Continuous textile headband ensures that the mask fits comfortably and securely
- Adjustable nose clip ensures an excellent individual fit and secure positioning
- Optional: exhalation valve to reduce heat and humidity in the mask
More sustainable mask family: the valve and headband attachment on all models are partly made of recycled material.
The soft material edges prevent uncomfortable pressure points. The seamless headband can be adjusted for an optimum fit. The exhalation valve with a 360° opening ensures very low breathing resistance and a cooling effect climate inside the mask. The upper face seal offers both a more secure positioning of the mask and increased wearer comfort.
Sealing lip (in the nose area for FFP1 and FFP2, all-round for FFP3) and nose clip provide maximum protection and comfort.
FFP3 preformed masks have a four-point headband attachment.
2220: additional activated carbon filter against odours, gases and vapours below threshold.
6 Options Available Below
Product Details & Documents
Respirators in protection class FFP1, FFP2 and FFP3.
- Soft material edges for greater comfort and wearer acceptance
- Continuous textile headband ensures that the mask fits comfortably and securely
- Adjustable nose clip ensures an excellent individual fit and secure positioning
- Optional: exhalation valve to reduce heat and humidity in the mask
More sustainable mask family: the valve and headband attachment on all models are partly made of recycled material.
The soft material edges prevent uncomfortable pressure points. The seamless headband can be adjusted for an optimum fit. The exhalation valve with a 360° opening ensures very low breathing resistance and a cooling effect climate inside the mask. The upper face seal offers both a more secure positioning of the mask and increased wearer comfort.
Sealing lip (in the nose area for FFP1 and FFP2, all-round for FFP3) and nose clip provide maximum protection and comfort.
FFP3 preformed masks have a four-point headband attachment.
2220: additional activated carbon filter against odours, gases and vapours below threshold.
Product Family Options
Product Information
- ClassWith valveTypeAvailabilityPrice
- UVEX8763100Folding mask, white
- UVEX8763110Folding mask, white
- UVEX8763200Folding mask, white
- UVEX8763210Folding mask, white
- UVEX8763220Folding mask, white
- UVEX8763310Folding mask, white