About this item
Loafer safety shoe incorporating MAXI-Soft Duo anti-fatigue technology designed for work stations that require standing for long periods (Upright or sustained activity).
- Recycled microfiber upper
- PU/PU sole Biomass Balance de BASF®
- ESD/Dissipative
- Polymer toe cap type B and midsole no metallic
- Inner sole in non-woven textile ESD
- Heel stiffener to reinforce the heel seat
Safety shoe protecting the electronic devices against electrostatic discharge phenomena. Electrostatic dissipative shoes ESD Environmental Class II tested according to EN 61340-4-3 and EN 61340-5-1.
Upper in recycled microfibre (40% recycled polyamide). Black colour.
Upper lined in breathable 3D Surf mesh (66% recycled polyester) with honeycomb structure to improve the peripheral ventilation of the foot, dries out rapidly.
Padded ergonomic collar in compact flexible foam, lined for extra comfort and better ankle and Achilles tendon protection.
Elasticated fastening for easy adjustment around the ankle joint.
Perforated insole for enhanced comfort : MAXI-Soft Duo Green composed of a soft Biomass Balance BASF® Dynamic PU midsole with heat sensitive memory foam and a maximum size Biomass Balance Elastopan insert that improves weight distribution by reducing pressure points and absorbs and relieves body stress associated with a prolonged standing working position and when walking. Anatomical, breathable and fitted with the Link ESDTM system, an innovative multi-contact, static electricity dissipater (Patent pending).
Ergonomic shoe, with broad synthetic polymer toe cap Springtane-B. Impact resistance to 200 Joules. Minimum height after test ≥4 mm in relation to the Type A toe cap in accordance with Standard 22568-2 : 2021.
BASF® PU/PU sole BMB - BioMass Balance Certifié EU-REDcert² with an urban sport design, linear profile and excellent slip resistance. Extremely light and flexible support base.