Uvex Sportstyle Planet Glasses
Supplier: UVEX
New Product
Certificates
About this item
Modern eye protection. Uvex sportstyle represents a new generation of innovative safety spectacles a clean, sporty design combining maximum functionality and protection, these lightweight safety glasses are a joy for everyone.
- Sporty, lightweight safety spectacles with contoured lens for a great fit on narrow or wider faces
- Uvex protecting planet - mass-balanced PC lens, frame and packaging partially made from recycled material
- Robust polycarbonate lens with proven uvex supravision coating technology - permanently anti-fog and scratch-resistant lens
- Detectable
- Frame colour: Beige, green/Anthracite, jade
- PC lens: Clear
- Universal fit
- UV protection
The combination of soft padding around the forehead and ergonomically shaped arms with soft ear-pieces ensures a comfortable and secure fit without creating pressure points. Soft, adjustable nose piece makes it possible to adjust the glasses to the wearer and ensures a comfortable, non-slip, pressure-free fit.
Single-lens glasses, innovative lens geometry, adjustable nose pad, soft nose piece, soft, non-slip sidearms.
Certifications: Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters). Certification: W 166 FT CE – 2C-1.2 W1 FTKN CE. UV400 – 100% protection against harmful UV radiation to 400 nm.