Order Entry
Germany
ContactUsLinkComponent
Human Recombinant SNX18 (from HEK293T)
Human Recombinant SNX18 (from HEK293T)
Catalog # ORIGTP319205
Supplier:  OriGene
New Product
CAS Number:  
Human Recombinant SNX18 (from HEK293T)
Catalog # ORIGTP319205
Supplier:  OriGene
New Product
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293T cells
  • Species:
    Human
  • Tag sequence:
    C-Myc/DDK
  • Protein synonyms:
    SNAG1|SH3PXD3B|SH3PX2
  • UniProtKB:
    Q96RF0
  • Protein/peptide name:
    SNX18
  • Purity:
    >80% as determined by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    68,8 kDa
  • Sequence:
    MALRARALYDFRSENPGEISLREHEVLSLCSEQDIEGWLEGVNSRGDRGLFPASYVQVIRAPEPGPAGDG
    GPGAPARYANVPPGGFEPLPVAPPASFKPPPDAFQALLQPQQAPPPSTFQPPGAGFPYGGGALQPSPQQL
    YGGYQASQGSDDDWDDEWDDSSTVADEPGALGSGAYPDLDGSSSAGVGAAGRYRLSTRSDLSLGSRGGSV
    PPQHHPSGPKSSATVSRNLNRFSTFVKSGGEAFVLGEASGFVKDGDKLCVVLGPYGPEWQENPYPFQCTI
    DDPTKQTKFKGMKSYISYKLVPTHTQVPVHRRYKHFDWLYARLAEKFPVISVPHLPEKQATGRFEEDFIS
    KRRKGLIWWMNHMASHPVLAQCDVFQHFLTCPSSTDEKAWKQGKRKAEKDEMVGANFFLTLSTPPAAALD
    LQEVESKIDGFKCFTKKMDDSALQLNHTANEFARKQVTGFKKEYQKVGQSFRGLSQAFELDQQAFSVGLN
    QAIAFTGDAYDAIGELFAEQPRQDLDPVMDLLALYQGHLANFPDIIHVQKGALTKVKESRRHVEEGKMEV
    QKADGIQDRCNTISFATLAEIHHFHQIRVRDFKSQMQHFLQQQIIFFQKVTQKLEEALHKYDSV
    TRTRPLEQKLISEEDLAANDILDYKDDDDKV
  • Concentration:
    >0,05 µg/µl as determined by microplate BCA method
  • Shipping temperature:
    Store at −80 °C

Specifications

About this item

Purified recombinant protein of Homo sapiens sorting nexin 18 (SNX18), transcript variant 1.

  • Cytogenetics: 5q11.2
  • Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.

Preparation: Recombinant protein was captured through anti-DDK affinity column followed by conventional chromatography steps.
Stability: Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.