Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Concentrators
Accessories for Vacuum concentrator, DNA SpeedVac system
Accessories for Vacuum concentrator, DNA SpeedVac system
THSAVLP200-230
:
THERMO SAVANT
Accessories for Vacuum concentrator, DNA SpeedVac system
THSAVLP200-230
:
THERMO SAVANT
Description:
Oil vacuum pump, 162 L/min throughput at 50 Hz, for use with DNA130-230, SPD120P2-230, SPD120P1-230
Pk:
1