Order Entry
Germany
ContactUsLinkComponent
 
Phenol red : ammonium sulphate (0.002 : 0.01) % in aqueous solution Reag. Ph. Eur. 1063604, buffered pH 4.7
Catalog # 87890.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Phenol red : ammonium sulphate (0.002 : 0.01) % in aqueous solution Reag. Ph. Eur. 1063604, buffered pH 4.7
Catalog # 87890.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:252-057-8
 :34487-61-1
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901