Order Entry
Germany
ContactUsLinkComponent
Ninhydrin solution R2 Reag. Ph. Eur. 1058305
Ninhydrin solution R2 Reag. Ph. Eur. 1058305
Catalog # 87878.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ninhydrin solution R2 Reag. Ph. Eur. 1058305
Catalog # 87878.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :178.14 g/mol
 :GHS05
Storage Temperature:Ambient
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791