Order Entry
Germany
ContactUsLinkComponent
Ninhydrin 1.7% in ethanol 83.3% (v/v) Reag. Ph. Eur. 1058304
Ninhydrin 1.7% in ethanol 83.3% (v/v) Reag. Ph. Eur. 1058304
Catalog # 87877.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ninhydrin 1.7% in ethanol 83.3% (v/v) Reag. Ph. Eur. 1058304
Catalog # 87877.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :178.14 g/mol
ADR:3,II
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791