Order Entry
Germany
ContactUsLinkComponent
1-Naphtholbenzeine 0.2% in acetic acid Reag. Ph. Eur. 1057601
1-Naphtholbenzeine 0.2% in acetic acid Reag. Ph. Eur. 1057601
Catalog # 87874.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
1-Naphtholbenzeine 0.2% in acetic acid Reag. Ph. Eur. 1057601
Catalog # 87874.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :374.44 g/mol
EINECS:205-656-3
UN:2789
ADR:8 (3),II
CAS Number:145-50-6
 :6948-88-5
Formula:C₂₇H₁₈O₂
MDL Number:MFCD00078492