Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Vaseline, white

Glass bottle for solids, 1 kg
Catalog # CSLO3200-1KG
Marketsource
Supplier:   CAESAR LORENTZ
Vaseline, white
Catalog # CSLO3200-1KG
Supplier:  CAESAR LORENTZ
Marketsource
CAS Number:  92045-74-4
Vaseline, white

Glass bottle for solids, 1 kg

CAS Number:  92045-74-4
Price
22,70 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Petrolatum, Petroleum Jelly
  • CAS Number:92045-74-4
  • Melting Pt:38…56 °C
  • Density:0.865 g/cm³ (25 °C)
  •  :Density: 0.815 to 0.88 g/cm³ (20 °C)
  • EINECS:295-456-2

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications