Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Celite® 545, filter aid
Celite® 545, filter aid
Celite® 545, filter aid
Health
Celite® 545, filter aid
Supplier:
Serva
Certificates
Expand image
Health
Celite® 545, filter aid
Supplier:
Serva
About this item
CAS Number:
68855-54-9
:
Formula:
SiO₂
MDL Number:
MFCD00132803
:
60.08 g/mol
EINECS:
272-489-0
:
GHS08