Potassium iodide 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1070502
Catalog # 87909.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:166 g/mol
EINECS:231-659-4
:GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405