Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Coomassie® Brilliant Blue R-250
Coomassie® brilliant blue R-250, Sigma-Aldrich®
Coomassie® brilliant blue R-250, Sigma-Aldrich®
Coomassie® brilliant blue R-250, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Coomassie® brilliant blue R-250, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
6104-59-2
:
Formula:
C₄₅H₄₄N₃NaO₇S₂
MDL Number:
MFCD00041762
:
825.98 g/mol
Melting Pt:
174…180 °C
Storage Temperature:
Ambient
EINECS:
228-060-5