Coomassie brilliant blue G-250, dark reddish purple powder C.I. 42655
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:6104-58-1
:
Formula:C₄₇H₄₈N₃NaO₇S₂
MDL Number:MFCD00078482
:854.04 g/mol
Density:>1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:228-058-4
Specification Test Results
- Identity Test:Passes
- UV/VIS: l max (methanol) :608 ± 3 nm